Promega Wizard™ Genomic DNA Purification Kits
An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
Supplier: Promega A1620
Includes: Cell lysis solution, nuclei lysis solution, protein precipitation solution, DNA rehydration solution, RNase solution‡
Description
- Manual, Solution-Based DNA Extraction Method
- DNA from blood, animal and plant cells, bacteria or yeast
- 60-minute, scalable protocol
- Kit size based on number of isolations from whole blood, other sample types may vary
† To fully rehydrate DNA, allow an additional hour at 65°C (149°F) or overnight at room temperature.
‡ PR-A1620 does not include RNase Solution.
Specifications
15°C to 30°C | |
3L Cell lysis solution, 1L Nuclei lysis solution, 350mL Protein precipitation solution, 150mL DNA rehydration solution | |
Genomic DNA Purification Kit | |
5 to 15μg/300μL |
Isolate genomic DNA from whole blood, cultured cells, yeast, mouse tissue or numerous other cell sources | |
100 Preps | |
10mL |