Promega Wizard™ MagneSil™ Sequencing Reaction Clean-Up System DFS Item
The Wizard™ MagneSil Sequencing Reaction Clean-Up System is designed for high-throughput purification of sequencing reactions, including BigDye Terminator reactions.
Supplier:  Promega A1831
 View more versions of this product
Catalog No. PRA1831
Explore available promotions
Includes: MagneSil GREEN Paramagnetic Particles and a protocol. MagneSil GREEN is also available separately.
Description
Description
  • High-Throughput Purification of DNA from Sanger Sequencing Reactions
  • Customized, walkaway methods possible
  • Purified products suitable for fluorescent sequencing
1 U.S. Patents 6,027,945 and 6,368,800 and Australian Patent 732756.
Specifications
Specifications
15°C to 30°C
Purifies sequencing reactions in a high-throughput system
Kit
Wizard™ Magnesil™ paramagnetic particle and a PCR plate
4 x 96 Preps