Promega Wizard™ SV 96 Plasmid DNA Purification System DFS Item
The Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format.
Supplier:  Promega A2250
 View more versions of this product
Catalog No. PR-A2250
Explore available promotions
Description
Description
  • Silica Membrane-Based, 96-Well, High-Throughput Plasmid Purification
  • 60-minute protocol
  • Manual and automated formats
  • Spin or vacuum-based methods
Specifications
Specifications
15°C to 30°C
Simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format
40mL Wizard™ SV 96 Cell resuspension solution, 40mL Wizard™ SV 96 Cell Lysis solution, 84mL Wizard™ SV 96 neutralization solution, 100mL Wizard™ SV 96 wash solution, 3mL Alkaline Prot
1 x 96 Preps