Promega Wizard™ SV Genomic DNA Purification System DFS Item
The Wizard™ SV Genomic DNA Purification System provides a fast, membrane-based method for preparing genomic DNA from cultured cells and tissue, including mouse tails.
Supplier:  Promega A2361
 View more versions of this product
Catalog No. PR-A2361
Explore available promotions
Includes:
  • Genomic DNA purification kit: Collection tubes, 0.5M EDTA (pH 8.0), Nuclease-free water, Nuclei lysis solution, RNase A solution, Wizard SV lysis buffer, Wizard SV minicolumns, Wizard SV wash solution
  • Genomic DNA Purification start-up kit (110V): Wizard SV Genomic DNA Purification System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold (20 sample capacity), vacuum adapters
Description
Description
  • Extracts DNA from Tissues or Cells
  • Isolate DNA from cultured cells in 20 minutes
  • Choice of spin or vacuum purification protocols
Specifications
Specifications
15°C to 30°C
Wizard™ SV minicolumns, collection tubes, 50mL nuclei Lysis solution, 30mL pH 8 0.5M EDTA, 2 x 50mL Wizard™ SV Lysis Buffer, 2 x 185mL Wizard™ SV wash solution, 2 x 1mL RNase A solution, 150mL Nuclease
System
Permits fast, simple preparation of genomic DNA without centrifugation of the lysate prior to purification
250 Preps