Learn More
Propargyl acrylate, 96%, stab. with ca 200ppm BHT, Thermo Scientific Chemicals
$158.29 - $158.29
Chemical Identifiers
CAS | 10477-47-1 |
---|---|
Molecular Formula | C6H6O2 |
Molecular Weight (g/mol) | 110.11 |
MDL Number | MFCD00078451 |
InChI Key | WPBNLDNIZUGLJL-UHFFFAOYSA-N |
Synonym | propargyl acrylate, 2-propenoic acid, 2-propynyl ester, acrylic acid propargyl ester, 2-propenoic acid, 2-propyn-1-yl ester, 2-propynyl acrylate, prop-2-yn-1-yl acrylate, acmc-1c257, prop-2-yn-1-yl prop-2-enoate, propargyl acrylate, |
PubChem CID | 82655 |
SMILES | C=CC(=O)OCC#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0938309
|
Thermo Scientific Chemicals
L0938309 |
10 g |
Each for $158.29
|
|
|||||
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
10477-47-1 | |
110.11 | |
WPBNLDNIZUGLJL-UHFFFAOYSA-N | |
82655 |
C6H6O2 | |
MFCD00078451 | |
propargyl acrylate, 2-propenoic acid, 2-propynyl ester, acrylic acid propargyl ester, 2-propenoic acid, 2-propyn-1-yl ester, 2-propynyl acrylate, prop-2-yn-1-yl acrylate, acmc-1c257, prop-2-yn-1-yl prop-2-enoate, propargyl acrylate, | |
C=CC(=O)OCC#C |
Specifications
10477-47-1 | |
138°C | |
Pungent | |
1.446 | |
MFCD00078451 | |
UN3272 | |
propargyl acrylate, 2-propenoic acid, 2-propynyl ester, acrylic acid propargyl ester, 2-propenoic acid, 2-propyn-1-yl ester, 2-propynyl acrylate, prop-2-yn-1-yl acrylate, acmc-1c257, prop-2-yn-1-yl prop-2-enoate, propargyl acrylate, | |
WPBNLDNIZUGLJL-UHFFFAOYSA-N | |
prop-2-ynyl prop-2-enoate | |
82655 | |
96% |
0.995 | |
43°C (109°F) | |
C6H6O2 | |
H2C=CHCO2CH2C≡CH | |
10 g | |
1902961 | |
Not miscible or difficult to mix with water. | |
C=CC(=O)OCC#C | |
110.11 | |
110.11 | |
Propargyl acrylate, stabilized with ≈200ppm BHT |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 233-975-8
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only