missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 131452500

 View more versions of this product

Catalog No. AC131452500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Propargyl alcohol
107-19-7,50-00-0
Colorless to Yellow
114.0°C to 115.0°C
Authentic
Glass bottle
1.4300 to 1.4330
MFCD00002912
01,454
0.949
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
TVDSBUOJIPERQY-UHFFFAOYSA-N
prop-2-yn-1-ol
7859
CHEBI:28905
99%
99%
-53.0°C
0.9490g/mL
34°C
98.5% min. (GC)
C3H4O
HC≡CCH2OH
250 mL
09,394
15,7923
Solubility in water: miscible. Other solubilities: miscible with benzene, chloroform, ethanol, ether,, 1,2-dichloroethane, acetone, dioxane, thf,pyridine, moderately soluble in ccl4, immiscible with aliphatic hydrocarbons
OCC#C
56.06
1.58 mPa.s (20°C)
56.06
Liquid

Chemical Identifiers

107-19-7
56.06
TVDSBUOJIPERQY-UHFFFAOYSA-N
7859
prop-2-yn-1-ol
C3H4O
MFCD00002912
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
CHEBI:28905
OCC#C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Suspected o

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 203-471-2

RTECSNumber : UK5075000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only