missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propargyl chloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 451840050

Catalog No. AC451840050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Propargyl chloride
1.0300g/mL
19°C
Glass bottle
HC≡CCH2Cl
5g
15,7924
Solubility in water: insoluble.
ClCC#C
74.51
74.51
624-65-7
58.0°C to 61.0°C
97.5% min. (GC)
C3H3Cl
MFCD00000980
1.03
propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
LJZPPWWHKPGCHS-UHFFFAOYSA-N
3-chloroprop-1-yne
12221
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call

GHS Signal Word: Danger

EINECSNumber : 210-856-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only