Learn More
Propionamide, 97%, Thermo Scientific Chemicals
$109.09 - $282.68
Chemical Identifiers
CAS | 79-05-0 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.09 |
MDL Number | MFCD00008039 |
InChI Key | QLNJFJADRCOGBJ-UHFFFAOYSA-N |
Synonym | propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47 |
PubChem CID | 6578 |
ChEBI | CHEBI:45422 |
IUPAC Name | propanamide |
SMILES | CCC(=O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC162670250
|
Thermo Scientific Chemicals
162670250 |
25 g | Plastic bottle |
Each for $109.09
|
|
||||
AC162671000
|
Thermo Scientific Chemicals
162671000 |
100 g | Plastic bottle |
Each for $282.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-05-0 | |
73.09 | |
QLNJFJADRCOGBJ-UHFFFAOYSA-N | |
6578 | |
propanamide |
C3H7NO | |
MFCD00008039 | |
propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47 | |
CHEBI:45422 | |
CCC(=O)N |
Specifications
79-05-0 | |
White to Yellow | |
>100°C | |
96% min. (GC) | |
C3H7NO | |
MFCD00008039 | |
02, 243 | |
propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47 | |
QLNJFJADRCOGBJ-UHFFFAOYSA-N | |
propanamide | |
6578 | |
73.09 | |
Crystalline Powder |
79.0°C to 83.0°C | |
213.0°C | |
Authentic | |
Plastic bottle | |
C2H5CONH2 | |
25 g | |
15, 7938 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, chloroform | |
CCC(=O)N | |
73.09 | |
CHEBI:45422 | |
97% | |
Propionamide, 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 201-172-1
RTECSNumber : UE2975000
TSCA : TSCA
RUO – Research Use Only