Learn More
Propionitrile, 99%, Thermo Scientific Chemicals
$105.18 - $268.45
Chemical Identifiers
CAS | 107-12-0 |
---|---|
Molecular Formula | C3H5N |
Molecular Weight (g/mol) | 55.08 |
MDL Number | MFCD00001948 |
InChI Key | FVSKHRXBFJPNKK-UHFFFAOYSA-N |
Synonym | propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril |
PubChem CID | 7854 |
ChEBI | CHEBI:26307 |
IUPAC Name | propanenitrile |
SMILES | CCC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA13203AK
|
Thermo Scientific Chemicals
A13203AK |
250 mL |
Each for $105.18
|
|
||||||
AAA13203AU
|
Thermo Scientific Chemicals
A13203AU |
1000 mL |
Each for $268.45
|
|
||||||
Description
Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.
Solubility
It decomposes with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Take measures to prevent the build up of electrostatic charge. Incompatible with Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
Chemical Identifiers
107-12-0 | |
55.08 | |
FVSKHRXBFJPNKK-UHFFFAOYSA-N | |
7854 | |
propanenitrile |
C3H5N | |
MFCD00001948 | |
propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril | |
CHEBI:26307 | |
CCC#N |
Specifications
107-12-0 | |
0.781 | |
6°C (42°F) | |
C3H5N | |
MFCD00001948 | |
UN2404 | |
14,7827 | |
It decomposes with water. | |
CCC#N | |
55.08 | |
CHEBI:26307 | |
99% |
-92°C | |
97°C to 98°C | |
Pleasant | |
1.366 | |
250 mL | |
773680 | |
propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril | |
FVSKHRXBFJPNKK-UHFFFAOYSA-N | |
propanenitrile | |
7854 | |
55.08 | |
Propionitrile |
Safety and Handling
GHS H Statement
H225-H300-H310-H331-H319
Highly flammable liquid and vapour.
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P330-P363-P370+P378q-P501c
H225-H300+H310-H319-H331
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPIONITRILE
EINECSNumber : 203-464-4
RTECSNumber : UF9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only