Learn More
Propiophenone, 99%, Thermo Scientific Chemicals
$50.59 - $407.23
Chemical Identifiers
CAS | 93-55-0 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.178 |
MDL Number | MFCD00009309 |
InChI Key | KRIOVPPHQSLHCZ-UHFFFAOYSA-N |
Synonym | propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one |
PubChem CID | 7148 |
ChEBI | CHEBI:425902 |
IUPAC Name | 1-phenylpropan-1-one |
SMILES | CCC(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1514022
|
Thermo Scientific Chemicals
A1514022 |
100 g |
Each for $50.59
|
|
||||||
AAA1514036
|
Thermo Scientific Chemicals
A1514036 |
500 g |
Each for $100.99
|
|
||||||
AAA151400E
|
Thermo Scientific Chemicals
A151400E |
2500 g |
Each for $407.23
|
|
||||||
Description
Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
93-55-0 | |
134.178 | |
KRIOVPPHQSLHCZ-UHFFFAOYSA-N | |
7148 | |
1-phenylpropan-1-one |
C9H10O | |
MFCD00009309 | |
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one | |
CHEBI:425902 | |
CCC(=O)C1=CC=CC=C1 |
Specifications
93-55-0 | |
1.008 | |
87°C (188°F) | |
1.526 | |
100 g | |
14,7830 | |
Difficult to mix. | |
CCC(=O)C1=CC=CC=C1 | |
134.178 | |
CHEBI:425902 | |
99% |
17°C to 18°C | |
217°C to 218°C | |
C9H10O | |
MFCD00009309 | |
606215 | |
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one | |
KRIOVPPHQSLHCZ-UHFFFAOYSA-N | |
1-phenylpropan-1-one | |
7148 | |
134.18 | |
Propiophenone |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
H227-H319
EINECSNumber : 202-257-6
RTECSNumber : UG7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only