Learn More
Propylamine, 98%, pure, Thermo Scientific Chemicals
$191.48 - $191.48
Chemical Identifiers
CAS | 107-10-8 |
---|---|
MDL Number | MFCD00008205 |
InChI Key | WGYKZJWCGVVSQN-UHFFFAOYSA-N |
Synonym | propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine |
PubChem CID | 7852 |
ChEBI | CHEBI:39870 |
IUPAC Name | propan-1-amine |
SMILES | CCCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC149310010
|
Thermo Scientific Chemicals
149310010 |
1 L | Glass Bottle |
Each for $191.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-10-8 | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
7852 | |
propan-1-amine |
MFCD00008205 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
CHEBI:39870 | |
CCCN |
Specifications
107-10-8 | |
Colorless to Yellow | |
48.0°C | |
Authentic | |
Glass Bottle | |
1.3870 to 1.3890 | |
MFCD00008205 | |
04,136 | |
0.719 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
propan-1-amine | |
7852 | |
CHEBI:39870 | |
98% | |
Liquid |
-83.0°C | |
0.7190g/mL | |
−30°C | |
97.5% min. (GC) | |
C3H9N | |
CH3CH2CH2NH2 | |
1 L | |
13,164 | |
15,7956 | |
Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether | |
CCCN | |
59.11 | |
0.4 mPa.s (20°C) | |
59.11 | |
Pure | |
Propylamine, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-462-3
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only