missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylamine, 99+%, extra pure, Thermo Scientific Chemicals

$28.68 - $105.70

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC220610500
SDS View Documents
Thermo Scientific Chemicals
220610500
50 mL Glass bottle
Each for $28.68
 
AC220615000
SDS View Documents
Thermo Scientific Chemicals
220615000
500 mL Glass bottle
Each for $74.30
 
AC220610010
SDS View Documents
Thermo Scientific Chemicals
220610010
1 L Glass bottle
Each for $105.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-10-8
59.11
WGYKZJWCGVVSQN-UHFFFAOYSA-N
7852
propan-1-amine
C3H9N
MFCD00008205
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
CHEBI:39870
CCCN

Specifications

107-10-8
0.7190g/mL
−30°C
99% min. (GC)
C3H9N
CH3CH2CH2NH2
50 mL
13,164
15,7956
Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether
CCCN
59.11
0.4 mPa.s (20°C)
59.11
Extra Pure
Propylamine, 99+%
-83.0°C
48.0°C
Authentic
Glass bottle
1.3870 to 1.3890
MFCD00008205
04,136
0.719
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propan-1-amine
7852
CHEBI:39870
99+%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-462-3

RTECSNumber : UH9100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only