missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylamine 98.0+%, TCI America™

Supplier:  TCI America P0520500ML

 View more versions of this product

Catalog No. P0520500ML

Explore available promotions

Description

Description

Specifications

Specifications

Propylamine
-83°C
12.6 (100g/L H2O 20°C)
C3H9N
500 mL
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
CCCN
59.112
CHEBI:39870
≥98.0% (GC,T)
107-10-8
Yellow
48°C
MFCD00008205
1277
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propan-1-amine
7852
59.11
Liquid

Chemical Identifiers

107-10-8
59.112
WGYKZJWCGVVSQN-UHFFFAOYSA-N
7852
propan-1-amine
C3H9N
MFCD00008205
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
CHEBI:39870
CCCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0131

RTECSNumber : UH9100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions