missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene, 98%

CAS: 103-65-1 | C9H12 | 120.20 g/mol

$47.80 - $142.04

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC418430250
SDS View Documents
Thermo Scientific Chemicals
418430250
25 g Glass bottle
Each for $47.80
Only null left
 
AC418431000
SDS View Documents
Thermo Scientific Chemicals
418431000
100 g Glass bottle
Each for $142.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-65-1
120.20
ODLMAHJVESYWTB-UHFFFAOYSA-N
7668
propylbenzene
C9H12
MFCD00009377
n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
CHEBI:42630
CCCC1=CC=CC=C1

Specifications

103-65-1
Yellow
158°C
Authentic
Glass bottle
1.4900 to 1.493
MFCD00009377
05, 390
15, 7957
Solubility in water: practically insoluble in water. Other solubilities: miscible with ethanol,ether and acetone
CCCC1=CC=CC=C1
120.20
CHEBI:42630
98%
Propylbenzene
-99°C
0.8600g/mL
47°C
97.5% min. (GC)
C9H12
C6H5CH2CH2CH3
25 g
0.86
n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
ODLMAHJVESYWTB-UHFFFAOYSA-N
propylbenzene
7668
120.19
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-132-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only