Learn More
Propylene carbonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1555230
Description
Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsPropylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.
Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
Specifications
Propylene Carbonate | |
°C to 49°C | |
240°C to 243°C | |
99% | |
1.421 | |
250 g | |
Miscible with water,alcohol,ether,acetone,benzene,chloroform and ethyl acetate. | |
CC1COC(=O)O1 | |
102.09 | |
102.09 |
108-32-7 | |
1.204 | |
132°C (270°F) | |
C4H6O3 | |
MFCD00005385,MFCD00798264,MFCD00798265 | |
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
4-methyl-1,3-dioxolan-2-one | |
7924 | |
Liquid |
Chemical Identifiers
108-32-7 | |
102.09 | |
RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
7924 | |
CC1COC(=O)O1 |
C4H6O3 | |
MFCD00005385,MFCD00798264,MFCD00798265 | |
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
4-methyl-1,3-dioxolan-2-one |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 203-572-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only