missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 57-55-6 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.095 |
MDL Number | MFCD00064272 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
IUPAC Name | propane-1,2-diol |
SMILES | CC(CO)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
P355-4
|
Thermo Fisher Scientific
P3554 |
4 L | Glass Bottle |
Each for $791.15
Case of 4 Each for $3,164.60
|
|
||||
Description
Propylene Glycol (USP/FCC)
Chemical Identifiers
57-55-6 | |
76.095 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
propane-1,2-diol |
C3H8O2 | |
MFCD00064272 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
Specifications
57-55-6 | |
-60°C | |
187°C | |
Glass Bottle | |
CH3CHOHCH2OH | |
4 L | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
propane-1,2-diol | |
1030 | |
76.1 | |
≥99.5% (USP) | |
Viscous Liquid |
Pass Test (FCC/USP) | |
6 to 8 | |
0.007% max. (FCC/USP) | |
C3H8O2 | |
MFCD00064272 | |
1.035 to 1.037 (FCC, USP) | |
Meets Requirements (USP) | |
CC(CO)O | |
76.095 | |
CHEBI:16997 | |
0.13mbar at 20°C | |
FCC/USP | |
Propylene Glycol |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:0
Flammability:1
Instability:0