Contec™ PROSAT™ Presaturated Nonwoven Wipes
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
Supplier:  Contec™ PSPP0043
 View more versions of this product
Catalog No. 19-033-773
Explore available promotions
Description
Description
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
  • Wipe substrate designed for the application
  • Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping articles prior to pass through
  • Made of meltblown polypropylene or cellulose/polyester
  • Effective and convenient for wipe-downs of process tools and other equipment
  • Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
  • Solvent blend meets SEMI specifications for metals and contaminants
Specifications
Specifications
Pre-saturated Wipe
28 cm
23 cm
Not Autoclavable
Single-use
For interior surfaces of process tools, surface preparation, manufacturing processes
50 Wipes/Bag, 40 Bags/Cs.
70% IPA, 30% DI Water
11 in.
11 in.
Polypropylene
ISO Class 5
28 x 23 cm (11 x 9 in.)
Process tools and other equipment
1-ply
Non-sterile