DuPont™ ProShield™ 50 Coveralls, Hood and Boots
Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.
Supplier:  DuPont™ NB122SWHXL002500
 View more versions of this product
Catalog No. 19-065-501D
Explore available promotions
Alert:
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Description
Description
  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Front zipper closure
  • Elastic wrists, boots
  • Attached hood
  • Serged seams
  • Collar
  • White
  • Thickness: 10 mils
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Attached Boots
Hooded
Front Zipper
Non-sterile
40.75 to 44.25 in.
70.48 cm
Janitorial, Sanitation, General Industrial, Maintenance
87.63 cm
68.58 cm
1.73 to 1.88 m
X-Large
Non-woven Fabric with Microporous Film
Single-use
Elastic
Standard, Elastic Face
Serged
NB122S
27.75 in.
10 mil
34.5 in.
27 in.
5.22 to 6.16 ft.
25/Cs.