missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ PROSHIELD™ 6SFR Coverall with Standard Fit Hood, Attached Socks
DuPont™ ProShield™ 6 SFR protective coveralls with standard fit hood and attached socks help extend the life of your flame resistant garments.
Supplier: DuPont™ TM122SBU4X002500
Description
DuPont™ ProShield® 6 SFR garments are cost-efficient and designed to be worn over primary flame resistant clothing.
- Keeps dirt and grime off of primary flame resistant garments
- Provides a barrier against non-hazardous particles and aerosols
Specifications
4X-Large | |
Blue | |
Attached Socks, Elastic | |
Hooded | |
Non-sterile | |
None |
Unisex | |
Single-use | |
Elastic | |
Serged | |
Flame, Particle | |
25/Cs. |