DuPont™ ProShield™ 10 Disposable SMS Frocks
Provides general protection against non-hazardous light liquid splashes and dry particulates.
Supplier:  DuPont™ PB271SWHMD003000
 View more versions of this product
Catalog No. 19-162-617
Explore available promotions
Description
Description
  • These limited-use garments have been optimized for comfort, softness, and breathability
  • Affordable and practical, ProShield™ garments can protect against non-hazardous dry particles and light liquid splash applications typically found in industrial situations
  • For non-hazardous, light duty jobs, DuPont™ has the protection your workers need
Specifications
Specifications
Medium
White
Elastic
None
SMS
Zipper
General Protection
39.75