DuPont™ ProShield™ 10 Disposable SMS Frocks
Provides general protection against non-hazardous light liquid splashes and dry particulates.
Supplier:  DuPont™ PB271SWH3X003000
 View more versions of this product
Catalog No. 19-162-654
Explore available promotions
Description
Description
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles in a choice of white, blue and gray. Applications include: general maintenance, janitorial/cleaning and other dirty work.
Compliance
Meets ASTM™ D1117 standards.
Specifications
Specifications
3X-Large
White
Front Zipper with Storm Flap
Serged
Coveralls, elastic wrists
SMS Fabric
Single-use
Elastic
General maintenance, janitorial/cleaning and other dirty work