Promega ProteaseMAX™ Surfactant, Trypsin Enhancer
ProteaseMAX Surfactant, Trypsin Enhancer, enhances the enzymatic performance of Trypsin, Chymotrypsin and Lys-C in digestion reactions.
Supplier: Promega V2072
Description
- Improved In-Gel and In-Solution Protein Digestion
- Enhances enzymatic performance of trypsin, chymotrypsin and Lys-C
- Saves time and labor when used with in-gel protein digestion
- Complete digestion in 1 hour
Specifications
Lyophilized | |
Trypsin Enhancer | |
Protein isolation and purification |
5 x 1mg ProteaseMAX™ Surfactant, lyophilized | |
-30°C to -10°C | |
5 x 1 mg |