missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Protector™ Benchtop Evidence Drying Cabinet
Provides a compact, space-saving method of securely drying evidence in a ventilated area
$13635.89 - $13743.04
Specifications
Cabinet Type | Benchtop evidence drying lockable cabinet |
---|---|
Depth (English) | 28 in. |
Depth (English) Interior | 26.5 in. |
Depth (English) Shelf | 19 in. |
Depth (Metric) | 71.1 cm |
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
10-359-159
|
Labconco™
3390001 |
220 V |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
10-359-158
|
Labconco™
3390000 |
110 V |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Lockable cabinet maintains the chain of custody. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA and carbon filters and is exhausted back to the room through the top of the cabinet. Filters protect personnel from exposure to biohazards while eliminating nuisance odors.
- Type 304 brushed stainless steel interior and epoxy-coated steel exterior
- Stainless-steel hanging rod
- Three stainless-steel collapsible/removable wire mesh shelves
- Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron
- Activated organic vapor carbon filter with 7.5 lb. (3.4 kg) porous coconut shell media
- Built-in 75 watt blower with solid state control rated for 5 amps
- Storage area: 26.5D x 36W x12H in. (67.3 x 91.4 x 30.5cm) with door and 14-quart plastic bucket
- Safety glass door with magnetic seal to accommodate user-supplied padlock or optional tamper-evident tag
- Key-activated ultraviolet light provides secondary disinfection of interior between cases
- 2 in. (50.8cm) diameter locking casters
- Inlet and exhaust roughing prefilters with MERV 11 rating
- Red filter system alarm light to alert operator to low airflow conditions
- Three-position switch: fan ON, OFF, ultraviolet light ON
- 96 in. (2.44m) three-wired cord and plug
- Dimensions: 28D x 36W x 45.7 in.H (71.1 x 91.4 x 116.1cm)
Order Info
Optional ducting to the outside is available.
Specifications
Benchtop evidence drying lockable cabinet | |
26.5 in. | |
71.1 cm | |
48.26 cm | |
HEPA and Carbon filter | |
12 in. | |
30.48 cm | |
UV light | |
Domestic | |
170 kg | |
36 in. | |
36 in. | |
91.4cm | |
91.44 cm | |
Evidence Drying Cabinet |
28 in. | |
19 in. | |
67.31 cm | |
Safety glass door with magnetic seal | |
45.7 in. | |
114.3 cm | |
30.48 cm | |
UV Light | |
375 lb. | |
36 in. | |
8 in. | |
91.4 cm | |
20.3 cm | |
Yes | |
Stainless Steel |