missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Protein A and Protein G Mag Sepharose™

Simplify enrichment of target proteins by immunoprecipitation or pull-down applications

$97.34 - $242.00

Specifications

Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Content And Storage 4°C to 8°C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Binding Capacity Ligand Bead Type Price Quantity  
Catalog Number Mfr. No. Quantity Binding Capacity Ligand Bead Type Price Quantity  
45-002-161
View Documents
Cytiva
28944006
500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
Each for $100.82
 
45-002-160
View Documents
Cytiva
28944008
500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
Each for $97.34
 
45-002-173
View Documents
Cytiva
28951379
4 x 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
Pack of 4 Each for $210.00
 
45-002-174
View Documents
Cytiva
28951378
4 x 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
Pack of 4 Each for $242.00
 
Description

Description

Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

  • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: Native protein A; Native protein G
Binding capacity Protein A: 8-17mg human IgG/mL gel
Binding capacity Protein G: 13-22mg human IgG/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% ethanol
Storage temperature: 4°C to 8°C

Specifications

Specifications

Paramagnetic spherical, highly cross linked agarose particles
37 to 100 μm
4°C to 8°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'