missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Protein G Sepharose™ 4 Fast Flow Media
Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.
$1033.11 - $8068.25
Specifications
Ligand Concentration | 2mg Protein G/mL Medium |
---|---|
Matrix | 4% Cross Linked Agarose |
Particle Size | 90 μm |
pH | 3 to 9 |
Content And Storage | 2°C to 8°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
45000116
|
Cytiva
17061801 |
5 mL |
Each for $1,033.11
|
|
|||||
45-000-117
|
Cytiva
17061802 |
25 mL |
Each for $3,498.51
|
|
|||||
45-000-118
|
Cytiva
17061805 |
200 mL |
Each for $8,068.25
|
|
|||||
Description
- Binding specificities that complement Protein A media
- Binds a broad range of IgG species and subclasses
- High capacity, >20mg human IgG/mL medium; high flowrate
- Multi-point attachment minimizes ligand leakage
- Used in a range of diagnostic and research applications
Specifications
2mg Protein G/mL Medium | |
90 μm | |
2°C to 8°C | |
Recombinant Protein G lacking albumin binding region |
4% Cross Linked Agarose | |
3 to 9 | |
Used in a range of diagnostic and research applications |