missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Protein Loading Buffer for SDS-PAGE, 5X

5X Protein Loading Buffer for SDS-PAGE is used for routine SDS-PAGE protein sample loading.

Supplier:  MP Biomedicals™ 08W00015

Catalog No. ICN08W00015

Explore available promotions

Description

Description

  • Modified, 5X concentrated protein loading buffer containing bromophenol blue dye and SDS
  • Without DTT
  • 5X protein loading dye for SDS-PAGE electrophoresis
  • Contains bromophenol blue dye
Specifications

Specifications

Protein Loading
5X
SDS-PAGE
1 mL
1 Vial
Blue
Electrophoresis
Loading Buffer
Liquid
-20°C
Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.