missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ Protein LoBind™ Deepwell Plates
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
Supplier: Eppendorf™ 951032921
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing in plates.
Specifications
Autoclavable | |
Yellow | |
Polypropylene | |
96 | |
Preparation or Storage of Protein, Peptide or Antibody Samples | |
PCR Clean (Non-sterile) | |
Colorless |
20 Plates (5 Bags x 4 Plates) | |
Protein LoBind™ Plate | |
1000 μL | |
Non-sterile | |
6000 x g | |
20 Pk. | |
85.5 x 127.8 x 44.1 mm |