missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Proteinase K (PK) Solution
Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride, 50% glycerol.
$998.75 - $998.75
Specifications
| Concentration | 20 mg/mL |
|---|---|
| pH | 4.3 to 12 |
| Product Type | Proteinase K (PK) Solution |
Description
- Stable at Room Temperature and Easy to Use
- Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
- Retains >80% activity at temperatures up to 60°C
- No resuspension or thawing before use
- Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml
Specifications
| 20 mg/mL | |
| 4.3 to 12 | |
| Proteinase K (PK) Solution |