missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PROTOCOL™ 10% Buffered Formalin, Fisher HealthCare™
Traditional fixative, buffered to a neutral pH
$41.91 - $1452.97
Chemical Identifiers
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
IUPAC Name | formaldehyde |
SMILES | C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
23-305510
|
Epredia
305510 |
32 oz. | Poly Bottle |
Each for $47.84
Case of 12 Each for $574.08
|
|
||||
23-245684
|
Epredia
245684 |
1 gal. | Poly Bottle |
Each for $41.91
Case of 4 Each for $167.64
|
|
||||
23-427098
|
Epredia
427098 |
2.5 gal. | Cubitainer |
Each for $121.72
|
|
||||
23-245685
|
Epredia
245685 |
5 gal. | Cubitainer |
Each for $120.00
|
|
||||
23-253998
|
Epredia
253998 |
55 gal. | Metal Drum |
Each for $1,452.97
|
|
||||
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
C=O |
CH2O | |
MFCD00003274 | |
formaldehyde |
Specifications
50-00-0 | |
32 oz. | |
MFCD00003274 | |
C=O | |
30.03 |
Poly Bottle | |
CH2O | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
Fisher PROTOCOL™ 10% Buffered Formalin |