missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PROTOCOL™ SafeFix™ II All-Purpose Fixative, Fisher HealthCare™
$585.16 - $585.16
Chemical Identifiers
CAS | 107-22-2 |
---|---|
Molecular Formula | C2H2O2 |
Molecular Weight (g/mol) | 58.036 |
InChI Key | LEQAOMBKQFMDFZ-UHFFFAOYSA-N |
Synonym | glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 |
PubChem CID | 7860 |
ChEBI | CHEBI:34779 |
IUPAC Name | oxaldehyde |
SMILES | C(=O)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
23-042601
|
Epredia
042601 |
5 gal. |
Each for $585.16
|
|
|||||
Chemical Identifiers
107-22-2 | |
58.036 | |
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
CHEBI:34779 | |
C(=O)C=O |
C2H2O2 | |
LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
7860 | |
oxaldehyde |
Specifications
107-22-2 | |
C2H2O2 | |
LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
oxaldehyde | |
7860 | |
PROTOCOL™ SafeFix™ II All-Purpose Fixative |
5 gal. | |
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
C(=O)C=O | |
58.036 | |
CHEBI:34779 |