missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ PTFE Tubing

For use in automatic analyzers, post column reactions, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds

Supplier:  MilliporeSigma™ Supelco™ 58700U

 View more versions of this product

Catalog No. 111016149


Explore available promotions

Description

Description

  • Maximum recommended operating temperature: 200°C
  • Short term exposure to higher temperatures seldom causes damage
Specifications

Specifications

Tubing
0.031 in.
10 ft.
500 psi (35 kg/cm2)
HPLC
1/16 in.
PTFE
200°C
Videos
SDS
Documents

Documents

Promotions

Promotions