Learn More
Thermo Scientific Chemicals PTP1B Inhibitor
Supplier: Thermo Scientific Chemicals J64512LB0
Description
PTP1B Inhibitor is used as a PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway. Protein tyrosine phosphatase 1B (PTP1B) has critical roles in cell signaling relevant to diabetes, obesity, and cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway
Specifications
5 mg | |
-30°C to -10°C | |
765317-72-4 | |
741.45g/mol | |
Light Sensitive | |
Signal Transduction Reagent | |
SXKBTDJJEQQEGE-UHFFFAOYSA-N | |
3-(3,5-dibromo-4-hydroxybenzoyl)-2-ethyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}-1-benzofuran-6-sulfonamide |
White | |
PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway | |
C26H19Br2N3O7S3 | |
3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethyl-N-[4-(2-thiazolylsulfamoyl)phenyl]benzofuran-6-sulfonamide | |
Soluble in ethanol and DMSO | |
Solid | |
CCC1=C(C(=O)C2=CC(Br)=C(O)C(Br)=C2)C2=CC=C(C=C2O1)S(=O)(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1 | |
741.44 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only