missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 390042500

 View more versions of this product

Catalog No. AC390042500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

250 g
Granular Stones
Plastic Bottle
C28H37FO7
CIWBQSYVNNPZIQ-YBDIZOPJSA-N
[2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate
76973482
Brown/Gray
Pumice stone
1332-09-8
MFCD00148147
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC
509.626

Chemical Identifiers

1332-09-8
509.626
CIWBQSYVNNPZIQ-YBDIZOPJSA-N
[2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate
C28H37FO7
MFCD00148147
76973482
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only