missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Pumice Stone, Honeywell Fluka™
Granulated
Chemical Identifiers
CAS |
1332-09-8 |
Molecular Formula |
C28H37FO7 |
Molecular Weight (g/mol) |
509.626 |
MDL Number |
MFCD00148147 |
InChI Key |
CIWBQSYVNNPZIQ-YBDIZOPJSA-N |
PubChem CID |
76973482 |
IUPAC Name |
[2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate |
SMILES |
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC |
View More Specs
Products
2
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
60044856
View Documents
|
Honeywell Chemicals
14372500G
|
500 g |
Poly Bottle |
|
|
Add to cart
|
|
|
|
60044855
View Documents
|
Honeywell Chemicals
143721KG
|
1 kg |
Poly Bottle |
|
|
Add to cart
|
|
|
|
Description
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Chemical Identifiers
|
1332-09-8 |
|
509.626 |
|
CIWBQSYVNNPZIQ-YBDIZOPJSA-N |
|
[2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate |
|
C28H37FO7 |
|
MFCD00148147 |
|
76973482 |
|
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC |
Specifications
|
500 g |
|
Granulated |
|
1332-09-8 |
|
MFCD00148147 |
|
CIWBQSYVNNPZIQ-YBDIZOPJSA-N |
|
[2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoate |
|
76973482 |
|
Gray |
|
Poly Bottle |
|
C28H37FO7 |
|
NONH for all modes of transport |
|
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC |
|
509.626 |
|
Pumice Stone |
Videos
SDS
missing translation for 'documents'