BUCHI Pure Essential Chromatography System
This system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
Supplier: BUCHI 11C10700
Description
- Benefits from a basic design and setup
- The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable
- Basic but powerful and safe thanks to a 50 bar (725 psi) pump, pressure sensors, and regulators
- Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900
- There are no limits regarding the size of the collected fractions
- Additional options allow you to be flexible regarding sample loading (liquid and solid) and the cartridge holder (any size between 4 – 5000 g)
- Intuitive navigation provided by novel and state-of-the-art software (methods, run data, and general settings)
- All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow
- Automation simplifies the entire separation process
Specifications
Pump: No; Detector: Yes; Fraction Collector: No | |
Flash Chromatography Kit I |