missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Biotix Pure uTIP™ Bulk Non-sterile Pipette Tips
Pure uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk are packaged in re-sealable bags making storage and racking simple. RNase, DNase, Pyrogen, and PCR Inhibitor free.
Supplier: Biotix 63305428
Description
- Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
- Packaged in re-sealable bags making storage and racking simple.
- Biotix™ uTIP™ Pure resin Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
- Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
- Achieved through patented technologies including FlexFit and Blade
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Re-sealable bag
- Tips must be racked by hand
Specifications
200 μL | |
Non-sterile | |
Autoclavable | |
Bulk, Bagged | |
1000 Pk. | |
Yellow | |
Universal Fit, Universal Fit Multichannel | |
uTip | |
PCR Ready |
Non-filtered | |
Clear | |
Universal | |
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point | |
Pipette Tips | |
Single-use | |
Yes | |
Polypropylene |
This product contains no safety hazards