missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gentex™ PureFlo ESM+ Visor Assembly DFS Item

Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.

Supplier:  Gentex™ PR02050SP

 View more versions of this product

Catalog No. 18444055

Explore available promotions

Description

Description

  • Spare visor assembly (Non Welding Unit)
  • For use with the PureFlo ESM+ PAPR
  • Color: Clear
Specifications

Specifications

Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.
Reusable
One Size
Yes
4/Pk.
Clear
Visor
PureFlo PF60 ESM+
PR02050SP
Non-sterile
Videos
SDS
Documents

Documents

Promotions

Promotions