missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PureLink™ RNase A (20 mg/mL)
PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA.
Supplier: Invitrogen™ 12091039
Includes: The PureLink 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A. Components are also available separately. PureLink 384 Plasmid Purification Plates are available in packs of 25. PureLink 384 Pre-Lysis Buffer is also available separately.
Description
- RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA
- Used for the isolation of RNA-free DNA
Concentration
- 20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA
Order Info
Shipping Condition: Room temperature
Specifications
1 x 25 mL bottle. Store at 2–8°C. |
|
Bottle | |
25 mL | |
Liquid |
20 mg/mL | |
UltraPure™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.