Learn More
Applied Biosystems™ PureQuant CD3+ T Cell Assay
The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples.
Supplier: Applied Biosystems™ A47197
Description
The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.
The PureQuant CD3+ Assay utilizes a specific demethylated site in the intragenic CD3G/CD3D region to positively identify CD3+ T-cells.
Benefits include:
- Proven performanceâcore technology has been used for identification and quantification of immune cell types for over a decade
- Easily standardizedâqPCR method enables consistent results across multiple users, instruments, and sites
- Facilitates regulatory filingsâregulatory support file provided upon request; established performance specifications following ICHQ2(R1) guidelines
- Scheduling flexibilityâassay can be performed using fresh or frozen cells/gDNA
The assays include all reagents, standards, and controls required to run the assay. Specific components include:
- Calibrator: corrects for variation in bisulfite conversion efficiency. Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
- Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
- Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.
This product is manufactured at a site with an ISO9001-certified quality management system.
Specifications
•Dynabeads SILANE Genomic DNA Kit, store at 2°C to 8°C • 1 x 2 mL PureQuant qPCR Master Mix II, store at 2°C to 8°C • 4 x ABS • THFA, store at room temperature • PureQuant CD3 Module, store at -15° to -25°C (CD3 and GAPDH TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA) |
|
Frozen | |
qPCR | |
QuantStudio™ | |
28 reactions | |
Cell |
Primer-probe | |
96-Well Plate | |
Fast or Standard | |
Purequant CD3 + T Cell Assay | |
28 reactions |
For Research Use Only. Not for use in diagnostic procedures.