missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Class I Safety Enclosure

Provide your staff with personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ Class I Safety Enclosure.

Supplier:  Labconco™ 3980203

 View more versions of this product

Catalog No. 10-304-111

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.

  • Total exhaust from 155 to 310cfm, depending on hood width
  • Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
  • Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
  • Airfoil with Clean-Sweep™ airflow openings
  • Optional built-in Guardian™ Airflow Monitor to alert user of face velocities outside preset range (where applicable)
  • With interlocked UV protection pane
  • Variable digital timer for UV lamp
  • Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
  • Class I Enclosures have a 15w, 254nm germicidal UV lamp to use in conjunction with surface disinfection to ensure decontamination of biohazardous particulates while unattended

  • 0.25 in. (6.4mm) Tempered glass side panels and sash
  • Glare-free fluorescent lamp mounted outside contamination area
  • Blower and light switches are front-mounted for easy access
  • Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
  • Back panel has two utility ports for service connections
  • Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface

For operator safety, UV lamp cannot be turned on when fluorescent lamp is in use.

Order Info

28.75L x 37 in. H (73 x 94cm). 115V 60Hz, 3A.

Certifications

ETL listed (certified to UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1).

Compliance

ASHRAE™ 110-1995 conformity tested. Biological containment verified per NSF Standard Number 49 Personnel Protection Test.

Specifications

Specifications

Class I Safety Enclosure
23.97 cm
With Airflow Monitor, UV Lamp
94.1 cm
23.75 in.
60 Hz
57 Kg
115 V
37.08 in.
Epoxy Coated Aluminum and Steel
Fluorescent, UV
110 to 155 cfm
60.32 cm
125 lb.
115 V, 60 Hz
3 A
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions