missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, US Models
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
$21539.93 - $36414.91
Specifications
Certifications/Compliance | NSF, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 |
---|---|
Chamber Material | Stainless Steel |
Product Type | Type A2 Biosafety Cabinet |
Model | Purifier Logic+ |
Noise Level | <63 dBA |
Catalog Number | Mfr. No. | Height (English) | Includes | Width (English) | Sash Opening (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Includes | Width (English) | Sash Opening (English) | Price | Quantity | ||
302310001
|
Labconco™
302310001 |
61.7 in. | Cabinet | 3 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302310101
|
Labconco™
302310101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 3 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302311001
|
Labconco™
302311001 |
61.7 in. | Cabinet with Accessory Package | 3 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302311101
|
Labconco™
302311101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 3 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302380001
|
Labconco™
302380001 |
61.7 in. | Cabinet | 3 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302380101
|
Labconco™
302380101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 3 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302381001
|
Labconco™
302381001 |
61.7 in. | Cabinet with Accessory Package | 3 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302381101
|
Labconco™
302381101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 3 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302410001
|
Labconco™
302410001 |
61.7 in. | Cabinet | 4 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302410101
|
Labconco™
302410101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 4 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302411001
|
Labconco™
302411001 |
61.7 in. | Cabinet with Accessory Package | 4 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302411101
|
Labconco™
302411101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 4 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302480001
|
Labconco™
302480001 |
61.7 in. | Cabinet | 4 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302480101
|
Labconco™
302480101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 4 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302481001
|
Labconco™
302481001 |
61.7 in. | Cabinet with Accessory Package | 4 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302481101
|
Labconco™
302481101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 4 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302510001
|
Labconco™
302510001 |
61.7 in. | Cabinet | 5 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302510101
|
Labconco™
302510101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 5 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302511001
|
Labconco™
302511001 |
61.7 in. | Cabinet with Accessory Package | 5 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302511101
|
Labconco™
302511101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 5 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302580001
|
Labconco™
302580001 |
61.7 in. | Cabinet | 5 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302580101
|
Labconco™
302580101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 5 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302581001
|
Labconco™
302581001 |
61.7 in. | Cabinet with Accessory Package | 5 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302581101
|
Labconco™
302581101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 5 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302610001
|
Labconco™
302610001 |
61.7 in. | Cabinet | 6 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302610101
|
Labconco™
302610101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 6 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302611001
|
Labconco™
302611001 |
61.7 in. | Cabinet with Accessory Package | 6 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302611101
|
Labconco™
302611101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 6 ft. | 10 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302680001
|
Labconco™
302680001 |
61.7 in. | Cabinet | 6 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302680101
|
Labconco™
302680101 |
89.2 to 95.2 in. | Cabinet, Base Stand | 6 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302681001
|
Labconco™
302681001 |
61.7 in. | Cabinet with Accessory Package | 6 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
302681101
|
Labconco™
302681101 |
89.2 to 95.2 in. | Cabinet with Accessory Package, Base Stand | 6 ft. | 8 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
All models feature:
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile (CAP) airflow monitoring system
- Intrinsically-safe negative pressure design
- Air-Wave™ Entry System
- Contain-Air™ Negative Pressure Channel
- Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
- Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
- Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
- Fully-closing, clear 1/4 in. tempered safety glass sash
- Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
- Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
- Smart-Start™ System user-programmable start up and shut down
- Night-Smart™ System reduces blower speed when sash is closed
- Leak-tight stainless steel interior and powder-coated steel exterior
- Removable towel catch located under work surface
- Code-activated electronic security lock
- 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
- 10° Angled sash with counterbalanced, easy lift mechanism
- ADA-Compliant touchpad control on right side post
- Noise level <63 dBA
- Heat load 990 BTU/hour (4 ft. models)
- Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
- Nominal inflow velocity of 105 fpm (0.5 m/sec)
- Nominal downflow velocity of 55 fpm (0.3 m/sec)
- Approximately 70% air recirculation
- Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
- 10 ft (3 m) power cord with plug
- Built-in timer and digital clock
- Five year warranty
- NSF International listed
- Optional NSF-approved non-welded telescoping base stand included on some models
- Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Specifications
NSF, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
Type A2 Biosafety Cabinet | |
<63 dBA | |
2 | |
60 Hz | |
79.248 cm |
Stainless Steel | |
Purifier Logic+ | |
4 | |
5 Years | |
31.2 in. | |
115 V |