Puritan™ HydraFlock™ Flocked Swabs
For collection and release of biological specimens
 View more versions of this product
Catalog No. 22-025-196
Explore available promotions
Description
Description
A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.; For buccal cell collection; Specimen collection; Tip material is comprised of polyester bicomponent fiber; Made in USA
Specifications
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
5.98 in.
Swab
15cm
14.9cm
1.6cm
0.21 in.
Polystyrene
Flocked
0.1 in.
Buccal cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection, DNA testing
0.254cm
5.89 in.
0.64 in.
0.55cm
Sterile
Large