missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ HydraFlock™ Sterile Flocked Swabs Packed in Dry Transport Tube

Packaged in a reclosable dry transport tube for ease in transport to the lab

Supplier:  Puritan™ 253606HBT

 View more versions of this product

Catalog No. 22-029-621


Explore available promotions

Description

Description

Puritan’s new dry transport system securely holds a swab in a round bottom tube made from virgin polypropylene resin, creating a safe and sterile environment to accommodate many different styles of collection devices. The injection molded cap assures a precision fit for multiple handle styles eliminating any foreign adhesive which may interfere with the specimen. Tubes include a distinctive label describing product identity, expiry date as well as allow for patient identification and collection information.

Certifications

CE marked (for medical use), ISO 9001:2008, ISO 13485:2003

Specifications

Specifications

CE, ISO 9001:2008, ISO 13485:2003
Buccal cell collection, forensic evidence collection, rapid diagnostic testing, DNA collection and testing, collection of Bacteria or microorganisms
152.4mm
30mm
17.3mm
0.21 in.
Dry transport tube
Polystyrene
6 in.
HydraFlock™ Flocked Swab
1.18 in.
0.68 in.
5.5mm
Medical
Sterile
Elongated flocked
SDS
Documents

Documents

Promotions

Promotions