missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puritan™ UniTranz-RT Transport Systems
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Supplier: Puritan™ UT117
Description
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USASpecifications
| UniTranz-RT Transport System | |
| Sterile | |
| CE | |
| Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma | |
| Individually-wrapped | |
| 300 Cs. | |
| 2°C to 25°C |
| Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt | |
| Single Applicator | |
| 1 mL | |
| High performance ultrafine purFlock ultra swab, wrapped and 1mL Universal Transport Medium Filled Vial | |
| 7.3 ±0.2 | |
| Polystyrene | |
| Ultrafine Flocked |