Puritan™ PurFlock™ Ultra Flocked Swabs
Ideal for rapid diagnostic tests
 View more versions of this product
Catalog No. 22-025-192
Explore available promotions
Description
Description
Elongated Flock Tipped Specimen Collection Applicator with Plastic Handle
PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultra’s high purity characteristics makes it an ideal swab for various rapid diagnostic tests.
Specifications
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2cm
15.2cm
Tapered
0.51cm
Molded break point
Polystyrene
Flocked
0.1 in.
For buccal cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection, DNA testing; molded breakpoint at 80 mm
0.254cm
6 in.
0.69 in.
1.72cm
0.2 in.
Sterile
Standard