Puritan™ PurFlock™ Ultra Flocked Swabs
Ideal for rapid diagnostic tests
 View more versions of this product
Catalog No. 22-025-193
Explore available promotions
Description
Description
Elongated Flock Tipped Specimen Collection Applicator with Plastic Handle
PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultra’s high purity characteristics makes it an ideal swab for various rapid diagnostic tests.
Specifications
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2cm
15.2cm
Tapered
0.34cm
Sterile
Minitip
0.098 in.
For Nasopharyngeal and Urethral cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection
0.24cm
6 in.
0.725 in.
1.8cm
0.135 in.
Polystyrene
Flocked