missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Puritan™ PurFlock™ Ultra Flocked Swabs
Ideal for rapid diagnostic tests
Description
Elongated Flock Tipped Specimen Collection Applicator with Plastic Handle
PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultraâs high purity characteristics makes it an ideal swab for various rapid diagnostic tests.Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's | |
6 in. | |
Swab | |
15.2cm | |
15.2cm | |
Tapered | |
0.35cm | |
Sterile | |
Ultrafine |
0.1 in. | |
For Nasopharyngeal and Urethral cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection | |
0.254cm | |
5.99 in. | |
0.66 in. | |
1.66cm | |
0.14 in. | |
Polystyrene | |
Flocked |