Learn More
MilliporeSigma™ Purospher™ STAR RP-18 Endcapped Hibar™ RT HPLC Columns, 3 micron
Fast method development of complex samples across the pH range from 1.5 to 10.5 at different mobile phases and temperature conditions
$1686.77 - $2798.17
Specifications
Particle Size 3 μm
Quantity 1/Pk.
Surface Area 330 m²/g
USP Type L1
Stationary Phase C18
View More Specs
Products 10
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
M1503980001
View Documents
MilliporeSigma™
1503980001
100 mm 3 mm
Each for $2,507.27
Only null left
M1504680001
View Documents
MilliporeSigma™
1504680001
250 mm 4 mm
Each for $2,798.17
Only null left
M1503930001
View Documents
MilliporeSigma™
1503930001
50 mm 3 mm
Each for $1,731.69
Only null left
M1504690001
View Documents
MilliporeSigma™
1504690001
100 mm 4.6 mm
Each for $2,323.01
Only null left
M1504270001
View Documents
MilliporeSigma™
1504270001
250 mm 3 mm
Each for $2,798.17
Only null left
M1504130001
View Documents
MilliporeSigma™
1504130001
125 mm 3 mm
Each for $2,323.01
Only null left
M1504700001
View Documents
MilliporeSigma™
1504700001
250 mm 4.6 mm
Each for $1,686.77
Only null left
M1504280001
View Documents
MilliporeSigma™
1504280001
50 mm 4 mm
Each for $1,761.97
Only null left
M1504140001
View Documents
MilliporeSigma™
1504140001
150 mm 3 mm
Each for $2,561.92
Only null left
M1504310001
View Documents
MilliporeSigma™
1504310001
125 mm 4 mm
Each for $2,398.93
Only null left
Description
Description
  • Highest silica purity (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and from batch-to-batch
  • No phase collapse when using highly aqueous mobile phases
  • Good suitability for LC-MS applications
  • The Hibar RT columns are complete with endfittings
Specifications
Specifications
3 μm
330 m²/g
C18
17%
Yes
Column
1/Pk.
L1
Silica
Reversed Phase
Reversed Phase
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'