missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heathrow Scientific™ Purple Reagent Reservoir Chiller
Reagent Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.
$242.65 - $242.65
Specifications
Color | Purple |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir Cooler |
Quantity | 2 Pk. |
Description
- Reagents are kept at less than 5°C (41°F) for approximately 3 hours
- Ice-free solution eliminates the risk of contamination from messy ice baths
- Stackable, save space when storing in the freezer or on the bench top
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
- Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
- Made from sonic welded, engineered polypropylene that ensures a leak free design
- Not intended for autoclave, do not use with liquid nitrogen
- Reusable
- Two-year warranty
Specifications
Purple | |
Non-sterile | |
2 Pk. | |
1 | |
Convoluted |
Polypropylene | |
Reservoir Cooler | |
Not Autoclavable | |
Reusable |