PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
$98.35 - $396.15
Specifications
For Use With (Application) | For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications |
---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness (English) | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness (English) | Quantity | Price | Quantity | ||
19-125-814
|
PURUS™
PS184538G |
Gray | 457 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $199.50
|
|
||
19-125-826
|
PURUS™
PS264538B |
Blue | 660 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $297.63
|
|
||
19-125-832
|
PURUS™
PS364538B |
Blue | 914 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $348.60
|
|
||
19-125-808
|
PURUS™
PS183638G |
Gray | 457 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $178.98
|
|
||
19-125-806
|
PURUS™
PS183638W |
White | 457 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $203.39
|
|
||
19-125-812
|
PURUS™
PS184538W |
White | 457 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $226.70
|
|
||
19-125-807
|
PURUS™
PS183638B |
Blue | 457 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $190.63
|
|
||
19-125-813
|
PURUS™
PS184538B |
Blue | 457 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $226.70
|
|
||
19-125-800
|
PURUS™
PS243034W |
White | 610 x 762 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $105.14
|
|
||
19-125-801
|
PURUS™
PS243034B |
Blue | 610 x 762 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $105.14
|
|
||
19-125-819
|
PURUS™
PS243638B |
Blue | 610 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $235.48
|
|
||
19-125-818
|
PURUS™
PS243638W |
White | 610 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $235.48
|
|
||
19-125-827
|
PURUS™
PS264538G |
Gray | 660 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $298.19
|
|
||
19-125-841
|
PURUS™
PS363638B |
Blue | 914 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $342.63
|
|
||
19-125-842
|
PURUS™
PS363638G |
Gray | 914 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $301.52
|
|
||
19-125-833
|
PURUS™
PS364538G |
Gray | 914 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $348.60
|
|
||
19-125-838
|
PURUS™
PM366034B |
Blue | 914 x 1524 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $278.34
|
|
||
19-125-840
|
PURUS™
PS363638W |
White | 914 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $342.63
|
|
||
19-125-839
|
PURUS™
PM366034G |
Gray | 914 x 1524 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $244.93
|
|
||
19-125-837
|
PURUS™
PM366034W |
White | 914 x 1524 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $261.34
|
|
||
19-125-860
|
PURUS™
PM183634W |
White | 457 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $108.90
|
|
||
19-125-831
|
PURUS™
PS364538W |
White | 914 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $396.15
|
|
||
19-125-862
|
PURUS™
PM363634B |
Blue | 914 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case for $160.97
|
|
||
19-125-861
|
PURUS™
PM183634B |
Blue | 18 x 36 in. (45.7 x 91.4 cm) | 1.4 mil | 4/Case |
Case of 4 Each for $102.25
|
|
||
19-125-825
|
PURUS™
PS264538W |
White | 660 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $295.08
|
|
||
17-125-730
|
PURUS™
PM364534G |
Gray | 914 x 1143 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $190.63
|
|
||
19-125-820
|
PURUS™
PS243638G |
Gray | 610 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 8 Mats/Cs. |
Case of 8 Each for $207.23
|
|
||
19-125-851
|
PURUS™
PS364538C |
Clear | 36 x 45 in. (91.4 x 114.3 cm) | 1.4 mil | 8/Case |
Case of 8 Each for $371.26
|
|
||
19-125-798
|
PURUS™
PM183634G |
Gray | 457 x 914 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $108.90
|
|
||
19-125-799
|
PURUS™
PM363634G |
Gray | 36 x 36 in. (91.4 x 91.4 cm) | 1.4 mil | 4/Case |
Case of 4 Each for $160.67
|
|
||
19-125-850
|
PURUS™
PM246034B |
Blue | 24 x 60 in. (61 x 152.4 cm) | 1.4 mil |
Case of 4 Each for $160.67
|
|
|||
19-125-950
|
PURUS™
PM264534B |
Blue | 26 x 45 in. (66 x 114.3 cm) | 1.4 mil |
Case of 4 Each for $155.97
|
|
|||
19-126-392
|
PURUS™
PM364534W |
White | 36 x 45 in. (91.4 x 114.3 cm) | 1.4 mil | 30 Sheets |
Case of 4 Each for $178.98
|
|
||
19-126-393
|
PURUS™
PM184534B |
Blue | 18 x 45 in. (45.7 x 114.3 cm) | 1.4 mil | 30 Sheets |
Case of 4 Each for $106.64
|
|
||
19-126-394
|
PURUS™
PM184534W |
White | 18 x 45 in. (45.7 x 114.3 cm) | 1.4 mil | 30 Sheets |
Case of 4 Each for $116.19
|
|
||
19-125-924
|
PURUS™
PS364538BHT |
Blue | 36 x 45 in. (91.4 x 114.3 cm) | 1.4 mil | 30 Layer |
Case of 8 Each for $348.60
|
|
||
19-125-802
|
PURUS™
PS243034G |
Gray | 610 x 762 mm | Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil | 30 Sheets/Mat, 4 Mats/Cs. |
Case of 4 Each for $98.35
|
|
||
19-125-965
|
PURUS™
PM243634G |
Gray | 24 x 36 in. (61 x 91.4 cm) | 1.4 mil |
Case of 4 Each for $132.50
|
|
|||
Description
- Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
- Strong & Effective delivering reliable and high-end performance
- Consistent adhesive coverage for maximum performance of each layer Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
- Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Certifications
Manufactured in ISO 9001 certified facility
Specifications
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications |